Learn More
Base/Neutrals Surrogate Standard Mix (For EPA 8270C)
Catalog No. USISM280N1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
Catalog No. Quantity
US-ISM-280N-1 1 x 1 mL
1 options
Catalog No. US-ISM-280N-1 Supplier Agilent Technologies Supplier No. ISM280N1
Each component at 100μg/mL in Methylene Chloride: Nitrobenzene-d5
Specifications
Chemical Name or Material Base/Neutrals Surrogate Standard Mix
Packaging Ampule
Quantity 1 x 1 mL