Learn More
Microsolv Technology Corporation Base Plate, Translucent Polypropylene, Micro-Sample Management System, universal, re-usable. 6/PK.
Catalog No. 502270530
Encompass_Preferred
Accommodates and holds one Micro-Sample Insert Rack in the U-2D System. Made of low weight polypropylene plastic, they comply with ANSI standards for 96 well plates. When used with Racks of Inserts, the unique molded bottoms allow each insert to be supported on the bottom. Use in a vortex mixer or water bath can be used with enhanced performance. Also most plate readers can be used. Bases are reusable and can be refrigerated.
This item is not returnable. View return policy
Catalog No. 50-227-0530 Supplier Microsolv Technology Corporation Supplier No. 95296SBPK
Accommodates and holds one Micro-Sample Insert Rack in the U-2D System. Made of low weight polypropylene plastic, they comply with ANSI standards for 96 well plates. When used with Racks of Inserts, the unique molded bottoms allow each insert to be supported on the bottom. Use in a vortex mixer or water bath can be used with enhanced performance. Also most plate readers can be used. Bases are reusable and can be refrigerated.