Learn More
Air Science Usa Base Stand 30" Wide for Ductless Fume Hoods
SDP
Catalog No. NC2684291
Encompass_Preferred
Base Stand, (Metal), Open Frame, Mobile, with Locking Casters, for 30" Nominal Width Models
Catalog No. NC2684291 Supplier Air Science Usa Supplier No. CART30
Base Stand, (Metal), Open Frame, Mobile, with Locking Casters, for 30" Nominal Width Models