Learn More
Air Science Usa Base Stand 48" Wide for Vertical Laminar Flow Cabinet
SDP
Catalog No. NC2684361
Encompass_Preferred
Base Stand, (Metal), Open Frame, Adjustable Leveling Feet, Fixed height. Fits Vlf-48 Models
Catalog No. NC2684361 Supplier Air Science Usa Supplier No. CARTBWVLF48
Base Stand, (Metal), Open Frame, Adjustable Leveling Feet, Fixed height. Fits Vlf-48 Models