Learn More
Air Science Usa Base Stand for Ductless Fume Hoods, Laminar Flow Cabinets, PCR Workstations, Vented Enclosures, Downflow Workstations, includes Locking Caster Wheels, 48" / 1200mm wide units
SDP
Catalog No. NC1792482
Encompass_Preferred
Base Stand (cart-50), Metal, Open Frame, Mobile, with Locking Casters, Lower Shelf, Fixed height, 28" Deep. Fits Purair Basic P5-48, Dws-48, Flow-48, PCR-48, Ve-48S, Ptefh-48, CA48S, and other 48" / 1200mm Nominal Width Models (cart-50)
Catalog No. NC1792482 Supplier Air Science Usa Supplier No. CART50
Base Stand (cart-50), Metal, Open Frame, Mobile, with Locking Casters, Lower Shelf, Fixed height, 28" Deep. Fits Purair Basic P5-48, Dws-48, Flow-48, PCR-48, Ve-48S, Ptefh-48, CA48S, and other 48" / 1200mm Nominal Width Models (cart-50)