Learn More
Ace Glass, Inc. BASE SYS,200L UNJACKETED "C"
SDP
Catalog No. 5000854797
Encompass
Unjacketed reactor base system designed for maximum diversity and ease of use. Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom designed parts. Each base system comes with the basic starting components required. Simply select the motor and accessories needed to complete the design. For customized components or application design, contact our technical department for further assistance.
Catalog No. 50-008-54797 Supplier Ace Glass, Inc. Supplier No. 1284623
Unjacketed reactor base system designed for maximum diversity and ease of use. Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom designed parts. Each base system comes with the basic starting components required. Simply select the motor and accessories needed to complete the design. For customized components or application design, contact our technical department for further assistance.