Learn More
Ace Glass, Inc. BASE SYSTEM,72L,SPH,UNJKTD "C"
SDP
Catalog No. 5000854759
Encompass
Unjacketed spherical reactor base system designed for maximum diversity and ease of use. Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom designed parts. Each base system comes with the basic starting components required. Simply select the motor and accessories needed to complete the design. For customized components or application design,contact our technical department for further assistance.
Catalog No. 50-008-54759 Supplier Ace Glass, Inc. Supplier No. 1284704
Unjacketed spherical reactor base system designed for maximum diversity and ease of use. Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom designed parts. Each base system comes with the basic starting components required. Simply select the motor and accessories needed to complete the design. For customized components or application design,contact our technical department for further assistance.