Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Base Ten Soft Interlocking Blocks
SCI_ED

Catalog No. S79417B
Change view
Click to view available options
Format:
1 x 1 x 1
1 x 1 x 10
1 x 10 x 10
10 x 10 x 10
Class Set
Starter Set
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S79417B 1 x 10 x 10
S79417D Starter Set
S79417F Class Set
S79417 1 x 1 x 1
S79417A 1 x 1 x 10
S79417C 10 x 10 x 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options

Choking Hazard – small parts.

Catalog No. S79417B Supplier Learning Resources, Inc Supplier No. LER 6354

Choking Hazard – small parts.

Includes

All sets include 96-page book and storage box.

Components conveniently interlock to teach math concepts while enabling students to easily manipulate and visualize varying quantities. Older students can explore spatial relationships and volume.

Starter set includes 100 units, 30 rods, 10 flats, and 1 cube. Place Value Set includes 100 units, 50 rods, 10 flats, 3 cubes, and 1 pad of Place Value charts (25 sheets). Class Set includes 600 units, 200 rods, 20 flats, and 3 cubes. Grades 1-6.

Specifications

Grade Level Elementary
Grade K to 2
Format 1 x 10 x 10
Product Type Interlocking Base Ten
Includes 10 Flats
For Use With (Application) Conveniently interlocks to clarify place value, estimation and operations concepts by enabling students to manipulate and visualize varying quantities with ease
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.