Learn More
Base Ten Soft Interlocking Blocks

Description
Includes
All sets include 96-page book and storage box.
Components conveniently interlock to teach math concepts while enabling students to easily manipulate and visualize varying quantities. Older students can explore spatial relationships and volume.
Starter set includes 100 units, 30 rods, 10 flats, and 1 cube. Place Value Set includes 100 units, 50 rods, 10 flats, 3 cubes, and 1 pad of Place Value charts (25 sheets). Class Set includes 600 units, 200 rods, 20 flats, and 3 cubes. Grades 1-6.
Specifications
Specifications
| Grade Level | Elementary |
| Grade | K to 2 |
| Format | 10 x 10 x 10 |
| Product Type | Interlocking Base Ten |
| Includes | 1 Cube |
| For Use With (Application) | Conveniently interlocks to clarify place value, estimation and operations concepts by enabling students to manipulate and visualize varying quantities with ease |
| Subject | Mathematics |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.