Learn More
Base Ten Soft Interlocking Blocks

Description
Includes
All sets include 96-page book and storage box.
Components conveniently interlock to teach math concepts while enabling students to easily manipulate and visualize varying quantities. Older students can explore spatial relationships and volume.
Starter set includes 100 units, 30 rods, 10 flats, and 1 cube. Place Value Set includes 100 units, 50 rods, 10 flats, 3 cubes, and 1 pad of Place Value charts (25 sheets). Class Set includes 600 units, 200 rods, 20 flats, and 3 cubes. Grades 1-6.
Specifications
Specifications
| Grade Level | Elementary |
| Grade | K to 2 |
| Format | Starter Set |
| Product Type | Interlocking Base Ten |
| Includes | 100 units, 30 rods, 10 flats, 1 cube and 96-page Base Ten Book |
| For Use With (Application) | Conveniently interlocks to clarify place value, estimation and operations concepts by enabling students to manipulate and visualize varying quantities with ease |
| Subject | Mathematics |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.