Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Baseball Bump Cap Head Protection

Catalog No. 19155463 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Black
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-463 -
19-155-462 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-155-463 Supplier Pyramex™ Supplier No. HP50031
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Baseball Bump Cap Head Protection offers a bump cap and baseball cap in one.

  • Reflective piping adds extra visibility
  • Soft inner foam cushion for comfortable wear
  • Size can be adjusted with a hook and loop strap to give a quick, secure fit
  • Tough polyethylene vented shell is removable and the outer cap is fully washable
  • Ideal for food processing and other industries that don't require full hard hat protection
  • Holes in the liner match up with two mesh panels on the cap to provide additional ventilation for coolness and comfort

Specifications

Certifications/Compliance Non-rated
For Use With (Application) Meter readers, food processors, and other industries that do not require full hard hat protection
Material Cotton, Polyester, and Inner shell of high impact acrylonitrile-butadiene-styrene
Product Type Bump Cap
Size 52 to 61 cm
Style Baseball Bump Cap
Ventilated Not Vented
Head Style Short Brim (Cap Style)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.