Learn More
Bioworld BASED, 100 mg
Supplier:  Bioworld 402000081
Encompass_Preferred
A homobifunctional, long chain cross-linking reagent. Variations of this compound have been used to prepare protein conjugates.PhotoreactiveThiol cleavableIodinatableAppearance: Solid
Catalog No. 50-255-381