Learn More
LGC GENOMICS LLC (Lucigen) Baseline-ZERO DNase, 5000 UDigest large or small DNA molecules better than standard DNase I.
Catalog No. NC1424104
Encompass
Baseline-ZERO™ DNase digests dsDNA and ssDNA into mononucleotides more effectively than the commonly used bovine pancreatic DNase I.
This item is not returnable. View return policy
Catalog No. NC1424104 Supplier LGC GENOMICS LLC Supplier No. DB0715K
Baseline-ZERO™ DNase digests dsDNA and ssDNA into mononucleotides more effectively than the commonly used bovine pancreatic DNase I.