Learn More
Newcomer Supply Basement Membrane, Gomori Stain Kit, 250 ml kit
SDP
Catalog No. NC2102288
Encompass
Newcomer Supply Basement Membrane, Gomori Stain Kit procedure, with included microwave modification, is a silver technique similar to the Jones Method for identification of glomerular and tubular basement membranes in renal tissue.
This item is not returnable. View return policy
Catalog No. NC2102288 Supplier Newcomer Supply Supplier No. 9167A
Newcomer Supply Basement Membrane, Gomori Stain Kit procedure, with included microwave modification, is a silver technique similar to the Jones Method for identification of glomerular and tubular basement membranes in renal tissue.