Learn More
Advanced Cell Diagnostics BaseScope™ VS Reagent Kit, BaseScope™ VS Assay is compatible on the Ventana Discovery Ultra IHC/ISH slide staining systems.
Catalog No. NC3682381
Encompass
BaseScope™ VS Reagent Kit is an RNA detection assay based on ACD's patented signal amplification and background suppression technology. This highly sensitive RNA ISH technique and can be used for short targets and splice variants. The BaseScope™ VS Reagent Kit is designed for use with the Discovery Ultra automated IHC/ISH slide staining systems by Ventana Medical Systems. This includes BaseScope™ VS Detection Reagents (323710), RNAscope Universal VS Sample Preparation Reagents v2 (PN 323740), and RNAscope VS Accessory Kit (320630). Each reagent kit provides enough reagents to stain ~60 slides
Catalog No. NC3682381 Supplier Advanced Cell Diagnostics Supplier No. 323700
Add to Cart
Add to Cart
Request a Quote
BaseScope™ VS Reagent Kit is an RNA detection assay based on ACD's patented signal amplification and background suppression technology. This highly sensitive RNA ISH technique and can be used for short targets and splice variants. The BaseScope™ VS Reagent Kit is designed for use with the Discovery Ultra automated IHC/ISH slide staining systems by Ventana Medical Systems. This includes BaseScope™ VS Detection Reagents (323710), RNAscope Universal VS Sample Preparation Reagents v2 (PN 323740), and RNAscope VS Accessory Kit (320630). Each reagent kit provides enough reagents to stain ~60 slides