Learn More
Zymo Research Corporation ZR BashingBead™ Lysis/Filtration Tubes w/ 0.5 mm Beads (50 ml) (25 Pack)
SDP
Catalog No. 501251736
Encompass_Preferred
The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tissues (2.0 mm) or microbes (0.5 mm). Used in the Quick-DNA Fungal/Bacterial Midiprep kit (Cat. #D6105).
Catalog No. 50-125-1736 Supplier Zymo Research Corporation Supplier No. S6010
The state of the art, ultra-high density beads are fracture resistant, chemically inert, and ideal for disrupting tissues (2.0 mm) or microbes (0.5 mm). Used in the Quick-DNA Fungal/Bacterial Midiprep kit (Cat. #D6105).