Learn More
Description
- It allows instructors to teach and practice CPR on patients with breasts, helping to eliminate misconceptions and ensuring that every trainee builds confidence and skill in Real-life cardiac emergencies.
- Enhance your BLS Training with the Basic Billy Female Chest Add-On
- The Add-on enables trainees to develop proper CPR hand placement and techniques using realistic female anatomy.
- Attaches easily to both BasicBilly and BasicBilly Plus versions, enabling quick setup and efficient training.
- Allows instructors to switch seamlessly between male and female chest skins for versatile CPR training.
- Designed for repeated classroom use, the product is durable, hygienic, and compatible with standard cleaning processes.
- The delivery content includes the Basic Billy Female Chest.
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | CPR Training |
| Color | Dark Skin |
| Gender | Female |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


