Learn More
Description
- Enables instructors to teach and practice CPR on patients with breasts, helping eliminate common misconceptions.
- Helps trainees build the confidence and skills needed to respond effectively in Real-life cardiac emergencies.
- Enhance your BLS Training with the Basic Billy Female Chest Add-On.
- Enables trainees to learn proper hand placement and technique on female anatomy.
- Attaches easily to both the BasicBilly and BasicBilly plus versions for quick setup and efficient training.
- Allows instructors to switch seamlessly between male and female chest skins.
- Ensures proper hand placement, realistic chest rise, and accurate compression performance.
- Designed for repeated classroom use with standard cleaning processes
- The delivery content includes the Basic Billy Female Chest.
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | CPR Training |
| Color | Light Skin |
| Gender | Female |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


