Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Basic Blood Collection Tray

Catalog No. 14127635 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-635 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-635
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blood collection tray tote to organize phlebotomy supplies.

  • Lightweight easy-to-clean ABS construction
  • Customize the compartments to suit your requirements with the dividers provided
  • Comfort grip on durable acetal plastic handle tested to hold up to 10 kg
  • Comes with a 17 mm well, 60-place, tube rack

Specifications

Material ABS
Product Type Phlebotomy Tray
Color White
Dimensions (L x W x H) 16.4 x 12.2 x 3.3 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.