Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basic Blue 1 90.0+%, TCI America™
SDP

Catalog No. P059825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P059825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P059825G Supplier TCI America Supplier No. P059825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3521-06-0
Molecular Formula C23H24Cl2N2
Molecular Weight (g/mol) 399.359
MDL Number MFCD00050260
InChI Key GRPFBMKYXAYEJM-UHFFFAOYSA-M
Synonym Primocyanine 6GX, Brilliant Basic Cyanine, Rhoduline Blue, Setoglaucine
PubChem CID 107404
IUPAC Name [4-[(2-chlorophenyl)-[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=CC=C3Cl.[Cl-]

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 399.36
Percent Purity ≥90.0% (HPLC)
Chemical Name or Material Basic Blue 1
EINECSNumber (5)-1989
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.