Learn More
Description
- Single Ply Meltblown Polypropylene construction
- Chemically inert, making it ideal for use with aggressive fluids
- Unique color helps to set these absorbents apart for proper disposal
- Perforated every 30 in. to help reduce waste
- Dimpled for added durability and reduced linting
Specifications
Specifications
| Color | Yellow |
| Material | Polypropylene |
| Type | Chemical Absorbent Pad |
| For Use With (Application) | Absorbent for applications with hazardous chemicals |
| Weight Capacity | Heavyweight |
| Dimensions (L x W) | 150 ft. x 30 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.