Learn More
Mabtech Inc ELISpot Flex: Human IFN-γ (ALP) kit, ELISpot Kit, Interferon gamma (IFN-γ), Human reactivity, ALP conjugate, For 4 plates, Ambient shipping temp, Storage 4-8 °C, Research use only
Supplier:  Mabtech Inc 34202A
Encompass
The MABTECH INC ELISpot Flex: Human IFN-γ (ALP) kit is designed to enumerate human IFN-γ secreting cells through the ELISpot assay. This versatile kit provides researchers with essential reagents for adaptable assay setups, allowing customization of plates, substrates, and antigens. It includes high-quality monoclonal capture and detection antibodies and Streptavidin-ALP for reliable detection. Suitable for use with peripheral blood mononuclear cells (PBMCs), it is intended for research purposes only.
  • Enables enumeration of cells secreting human IFN-γ
  • Includes matched capture and detection antibodies
  • Flexible assay setup with various plate and antigen options
  • Validated for peripheral blood mononuclear cells (PBMCs)
  • For research use only
Catalog No. NC1420496