Learn More
Creative Biomart Basic Fibroblast Growth Factor (FGF-b) Protein
Catalog No. 502084287
Encompass_Preferred
Fibroblast Growth Factor-2 Human Recombinant (FGF-2) produced in E. Coli is a single, non-glycosylated, polypeptide chain containing 154 amino acids and having a molecular mass of 17.2kDa. The FGF-b is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4287 Supplier Creative Biomart Supplier No. THP0236
Fibroblast Growth Factor-2 Human Recombinant (FGF-2) produced in E. Coli is a single, non-glycosylated, polypeptide chain containing 154 amino acids and having a molecular mass of 17.2kDa. The FGF-b is purified by proprietary chromatographic techniques.