Learn More
Electron Microscopy Sciences Basic Fuchsin 0.5% 250 ML
SDP
Catalog No. 5031857
Encompass_Preferred
Used in Lillie's Method for the staining of ferric and ferrous iron. See Lillie, R.D., Histopathologic Technic & Practical Histo-Chemistry, 3rd Ed., McGraw-Hill, N.Y., p. 407, c.1965 or Luna, G., (ed) Manual of Histologic Staining Methods of the AFIP, 3rd Ed., McGraw-Hill, N.Y., p. 182, c. 1968.
Catalog No. 50-318-57 Supplier Electron Microscopy Sciences Supplier No. 2660403
Used in Lillie's Method for the staining of ferric and ferrous iron. See Lillie, R.D., Histopathologic Technic & Practical Histo-Chemistry, 3rd Ed., McGraw-Hill, N.Y., p. 407, c.1965 or Luna, G., (ed) Manual of Histologic Staining Methods of the AFIP, 3rd Ed., McGraw-Hill, N.Y., p. 182, c. 1968.