Learn More
Bioworld Basic Fuchsin, 5 g
Catalog No. 50199189
Encompass_Preferred
A metachromatic stain of cytoplasm and nuclei. High Purity Grade Solubility test: Passes EM (543nm, 50% methanol): 86,900 Dye content >88% A550 >0.65 Staining, leilson procedure: Passes
Catalog No. 50-199-189 Supplier Bioworld Supplier No. 217700061
A metachromatic stain of cytoplasm and nuclei. High Purity Grade Solubility test: Passes EM (543nm, 50% methanol): 86,900 Dye content >88% A550 >0.65 Staining, leilson procedure: Passes