Learn More
Electron Microscopy Sciences Basic Fuchsin, Certified, C.I. DcFb-26 25 GR
SDP
Catalog No. 50980195
Encompass_Preferred
CAS #632-99-5 C₂₀H₁₉N₃·HCl
Basic Fuchsin is a rapid polychrome stain for epoxy-embedded tissue. Berkowitz et al., (1968). Selective staining of nervous tissue for light microscopy following preparation for electron microscopy. J. Histochem. Cytochem. 16:808. Sato, T., and Shamoto, M (1973). A simple rapid polychrome stain for epoxy-embedded tissue. Stain Technol. 48:223.
Catalog No. 50-980-195 Supplier Electron Microscopy Sciences Supplier No. 11260
CAS #632-99-5 C₂₀H₁₉N₃·HCl
Basic Fuchsin is a rapid polychrome stain for epoxy-embedded tissue. Berkowitz et al., (1968). Selective staining of nervous tissue for light microscopy following preparation for electron microscopy. J. Histochem. Cytochem. 16:808. Sato, T., and Shamoto, M (1973). A simple rapid polychrome stain for epoxy-embedded tissue. Stain Technol. 48:223.