Learn More
Abcam Basic fuchsin hydrochloride, Acid-fast stain, 25G
Catalog No. 502704708
Encompass_Preferred
MW 341.52 g/mol. Mixture of pararosaniline acetate and pararosaniline hydrochloride used for acid-fast staining.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4708 Supplier Abcam Supplier No. AB14633525G
MW 341.52 g/mol. Mixture of pararosaniline acetate and pararosaniline hydrochloride used for acid-fast staining.
The product is subject to the following: Abcam Restricted Use Statement