Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basic Fuchsin, Sigma-Aldrich

Catalog No. 01258923 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-923 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-923 Supplier MilliporeSigma Supelco Supplier No. B090425G
Add to Cart
Add to Cart

Basic fuchsin possesses anesthetic, bactericidal (gram-positive), and fungicidal properties.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Basic Fuchsin
Grade Certified
Color Black to Green
pH 5 to 6
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.