Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basic Fuchsin, Spectrum™ Chemical
SDP

Catalog No. 18600853 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-853 5 g Amber Glass Bottle
18-600-854 25 g Amber Glass Bottle
18-600-855 100 g Amber Glass Bottle
18-600-856 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-853 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BA1305GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

BA130, 569-61-9, C19H17N3·HCl

Spectrum™ Chemical Basic Fuchsin is a magenta dye that is a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin is a dark green crystal often used in the laboratory where it is employed to stain bacteria and in the fabrication of Schiff reagent. It is made up of a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin also has uses in the textile industry, and has many other names, including Basic Parafuchsin, Basic Red 9, Paramagenta Hydrochloride, and Pararosaniline Chloride.

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.82
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
SMILES [H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Grade Reagent
Physical Form Crystalline Powder or Lumps
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.