Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Basic Green 4, Certified, C.I. DcMg-11 25 GR
SDP

Catalog No. 5024624
Encompass_Preferred

(Malachite Green Oxalate; Diamond Green B; BX or P Extra; Light Green N, Solid Green O; Victoria Green B or WB)
CAS #2437-29-8 (C₂₃H₂₅N₂)₂.(C₂H₂O₄)₂.C₂O₄ F.W. 927.03
Solubility: 10.0% Water; 8.5% Alcohol; 5.5% Cellosolve; 7.0% Glycol; 0.0% Xylene

Used as a bacteria stain and a counterstain for botanical material.

Catalog No. 50-246-24 Supplier Electron Microscopy Sciences Supplier No. 18100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Malachite Green Oxalate; Diamond Green B; BX or P Extra; Light Green N, Solid Green O; Victoria Green B or WB)
CAS #2437-29-8 (C₂₃H₂₅N₂)₂.(C₂H₂O₄)₂.C₂O₄ F.W. 927.03
Solubility: 10.0% Water; 8.5% Alcohol; 5.5% Cellosolve; 7.0% Glycol; 0.0% Xylene

Used as a bacteria stain and a counterstain for botanical material.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.