Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Basic Life Support Simulator, BASICBilly+
SDPSCI_ED

Catalog No. S27925 Shop All 3B Scientific Products
Change view
Click to view available options
Color:
Dark Skin
Light Skin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S27925 Dark Skin
S27924 Light Skin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27925 Supplier 3B Scientific™ Supplier No. 8000953

missing translation for 'validMainframeMsg'

Includes a direct CPR Feedback Upgrade Kit which enables students and instructors to effectively train CPR with free, accurate, and user friendly mobile apps powered by heartisenseTM

Solid and Compact Manikin for Effective Training:

  • Made of easy to clean durable, latex free material
  • Hygienic, with easy to change disposable airways
  • Realistic anatomical landmarks to learn correct hand position for chest compression
  • Mouth-to-mouth, mouth-to-nose or BVM resuscitation can be practiced
  • Offers head-tilt functionality to open the airway or simulate obstructed airway
  • Visible chest rise when the lungs are ventilated
  • Force required for effective chest compressions has been carefully studied to offer a realistic training approach
  • Operation requires (2) AA batteries, not included

The CPR Apps Provide Feedback For:

  • Compression (depth, recoil, and rate)
  • Breath (volume)
  • Hands-off time in real-time
  • Feedback and evaluation meet the standard of the latest guidelines of the American Heart Association (AHA) and the European Resuscitation Council (ERC)

Overview of the CPR App Features:

  • Offers overall scoring and intuitive point of improvement during CPR
  • Direct (real-time) and summative CPR feedback for students
  • Instructor view can monitor up to 6 manikins at a time
  • Saved student results for review
  • Export or share students result/certificate as a PDF file
  • Instructor can use CPR analysis for detailed debriefing
  • Easily switch from conventional CPR training to Compression Only CPR settings
  • Stable connection is provided by a reliable Bluetooth connection
  • Mobile devices used to access the apps are not included in the delivery scope
  • An Upgrade Kit (1022626) is available to equip previously purchased BASICBilly manikins with the AHA required direct feedback functionalities

Specifications

Product Type AHA Compliant, BLS Manikin
For Use With (Application) Train, Monitor, Analyze, and Debrief CPR Performances
Color Dark Skin
Includes BASICBilly BLS Manikin (1), Adult Lung Bags (6), Adult Cardiac Massage Spring (1), Face Mask with Insert and Lung Bag Holder (1), Carrying Bag with Training Mat (1), BASICBilly+ Upgrade Kit (1), Add-On Kit with Breath Module Sensor (1), ColorID Stickers (1), Installation Manual (1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.