Learn More
Lonza Walkersville Basic Nucleofector Kit for Primary Mammalian Neurons (SCN)
Supplier: Lonza Walkersville VSPI1003
This is a neural cell specific Basic Nucleofector Kit which allows to determine the optimal transfection conditions for any mammalian neuron type. In contrast to our standard Basic Kit (VPI-1003) this kit is suited for transfection of small cell numbers.IMPORTANT NOTE: It only works in combination with an older Nucleofector Device model, the Nucleofector IIS Device.Benefits:Efficient transfection of neural or glial cells in small scale (20 µL)Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Applications:Transfection of low cell numbers in the range of 2 - 5 x10⁴ cellsKit suited for various neuron types from different mammalian species