Learn More
Lonza Walkersville Basic Nucleofector Kit for Primary Mammalian Neurons
Catalog No. NC9700172
Encompass
In addition to various cell type-specific kits for primary neurons, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian neuron type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for neurons. Benefits:Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 92%Viabilities up to 80%Applications:Kit suited for various neuron types from different mammalian species
Catalog No. NC9700172 Supplier Lonza Walkersville Supplier No. VPI1003
In addition to various cell type-specific kits for primary neurons, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian neuron type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for neurons. Benefits:Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 92%Viabilities up to 80%Applications:Kit suited for various neuron types from different mammalian species