Learn More
Lonza Walkersville Basic Nucleofector Kit for Primary Mammalian Fibroblasts
Catalog No. NC9443165
Encompass
In addition to various cell type-specific kits for primary fibroblasts, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian fibroblast type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for fibroblasts.Benefits:Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 90%Viabilities up to 98%Applications:Kit suited for fibroblasts from different mammalian species and various organs Already tested for macaque dermal fibroblasts bovine fibroblasts human colon myofibroblasts mouse lung fibroblasts etc.
Catalog No. NC9443165 Supplier Lonza Walkersville Supplier No. VPI1002
In addition to various cell type-specific kits for primary fibroblasts, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian fibroblast type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for fibroblasts.Benefits:Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 90%Viabilities up to 98%Applications:Kit suited for fibroblasts from different mammalian species and various organs Already tested for macaque dermal fibroblasts bovine fibroblasts human colon myofibroblasts mouse lung fibroblasts etc.