Learn More
Lonza Walkersville Basic Nucleofector Kit for Primary Mammalian Epithelial Cells
Catalog No. NC9954220
Encompass
In addition to various cell type-specific kits for primary epithelial cells, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian epithelial cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II).Benefits:Optimization can be performed within 1 experiment Detailed protocol guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 83%Viabilities up to 98%Applications:Kit suited for epithelial cells from different mammalian species and various organsAlready tested for renal proximal tubular epithelial cells (RPTEC) human prostate epithelial cells (hPrEc) and human small airway epithelial cells (SAEC)
Catalog No. NC9954220 Supplier Lonza Walkersville Supplier No. VPI1005
In addition to various cell type-specific kits for primary epithelial cells, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian epithelial cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II).Benefits:Optimization can be performed within 1 experiment Detailed protocol guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 83%Viabilities up to 98%Applications:Kit suited for epithelial cells from different mammalian species and various organsAlready tested for renal proximal tubular epithelial cells (RPTEC) human prostate epithelial cells (hPrEc) and human small airway epithelial cells (SAEC)