Learn More
Lonza Walkersville Basic Nucleofector Kit for Primary Mammalian Endothelial Cells
Catalog No. NC9498181
Encompass
In addition to various cell type-specific kits for primary endothelial cells, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian endothelial cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for endothelial cells. Benefits:Optimization can be performed within 1 experiment Detailed protocol guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 90%Viabilities up to 85%Applications:Kit suited for endothelial cells from different mammalian species and various organsAlready tested for human pulmonary artery endothelial cells
Catalog No. NC9498181 Supplier Lonza Walkersville Supplier No. VPI1001
In addition to various cell type-specific kits for primary endothelial cells, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian endothelial cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for endothelial cells. Benefits:Optimization can be performed within 1 experiment Detailed protocol guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support Team Transfection efficiencies up to 90%Viabilities up to 85%Applications:Kit suited for endothelial cells from different mammalian species and various organsAlready tested for human pulmonary artery endothelial cells