Learn More
Harrick Plasma Basic Plasma Cleaner 115V
SDP
Catalog No. NC9332171
Encompass_Preferred
The PDC-32G Basic Plasma Cleaner is our most compact, inexpensive benchtop plasma cleaner, suitable for nanoscale surface cleaning and surface chemistry modification of small samples. Maximum RF power of 18W; Adjustable RF power settings (LOW, MEDIUM, HIGH); 3in. diameter x 6.5in. length Pyrex chamber; Hinged door with viewing window; Active fan cooling; Integral switch for a vacuum pump; Metering valve to qualitatively control gas flow and chamber pressure; Three-way valve to quickly switch from introducing gas, venting, and isolating the chamber; Size: 8.5in. H x 10in. W x 8in. D; Weight: 13 lb. REQUIRED: A separate vacuum pump (minimum pump speed 23 L/min, ultimate total pressure of 200 mTorr or less) is required to run the plasma process.
This item is not returnable. View return policy
Catalog No. NC9332171 Supplier Harrick Plasma Supplier No. PDC32G
Add to Cart
Add to Cart
Request a Quote
The PDC-32G Basic Plasma Cleaner is our most compact, inexpensive benchtop plasma cleaner, suitable for nanoscale surface cleaning and surface chemistry modification of small samples. Maximum RF power of 18W; Adjustable RF power settings (LOW, MEDIUM, HIGH); 3in. diameter x 6.5in. length Pyrex chamber; Hinged door with viewing window; Active fan cooling; Integral switch for a vacuum pump; Metering valve to qualitatively control gas flow and chamber pressure; Three-way valve to quickly switch from introducing gas, venting, and isolating the chamber; Size: 8.5in. H x 10in. W x 8in. D; Weight: 13 lb. REQUIRED: A separate vacuum pump (minimum pump speed 23 L/min, ultimate total pressure of 200 mTorr or less) is required to run the plasma process.