Learn More
AdipoGen Basic Red 12
SDP
Catalog No. 502561611
Encompass_Preferred
Chemical. CAS 6320-14-5. Formula C25H29ClN2. MW 392.97. Basic red 12 magento is a cationic dye, a class of synthetic dyes acting as bases. These basic dyes are widely used in the textile industries, acrylic fibers manufacturing and many other industrial applications, such as printing on paper, flexo inks, crayons and dyeing on fibers, textiles, silk and wool. Spectral data The steady-state fluorescence maximum for an aqueous solution is at 560nm RT.
Catalog No. 50-256-1611 Supplier AdipoGen Supplier No. CDXB0279G005
Chemical. CAS 6320-14-5. Formula C25H29ClN2. MW 392.97. Basic red 12 magento is a cationic dye, a class of synthetic dyes acting as bases. These basic dyes are widely used in the textile industries, acrylic fibers manufacturing and many other industrial applications, such as printing on paper, flexo inks, crayons and dyeing on fibers, textiles, silk and wool. Spectral data The steady-state fluorescence maximum for an aqueous solution is at 560nm RT.