Learn More
Specac BASIC SOLIDS PACK
Catalog No. 501350583
Encompass_Preferred
Basic Solid Pack (for 7mm diameter KBr pellet preparation): Contains Mini-pellet press (2T), 7mm pellet die with pellet ring holder, 7mm spare pellet ring holder, pestle and mortar, KBr powder (50g), 7mm Disc holder with Rect. Mount.
Catalog No. 50-135-0583 Supplier Specac Supplier No. GS01150
Basic Solid Pack (for 7mm diameter KBr pellet preparation): Contains Mini-pellet press (2T), 7mm pellet die with pellet ring holder, 7mm spare pellet ring holder, pestle and mortar, KBr powder (50g), 7mm Disc holder with Rect. Mount.