Learn More
Fisherbrand™ Basic Steel Work Bench Complete Plumbing Assembly
Catalog No. 06000678 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Color Coded Brown
Color Coded Orange
Color Coded Yellow
Product Type:
Air Service Complete Plumbing Assembly
Nitrogen Service Complete Plumbing Assembly
Vacuum Service Complete Plumbing Assembly
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
06-000-678 Color Coded Yellow Nitrogen Service Complete Plumbing Assembly
06-000-677 Color Coded Orange Air Service Complete Plumbing Assembly
06-000-679 Color Coded Brown Vacuum Service Complete Plumbing Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-000-678
Add to Cart
Add to Cart
Fisherbrand™ Plumbing Assemblies are to be used with Fisherbrand Basic Work Bench.
  • Plumbing assemblies include a fine control needle valve service turret with shank, a service hose, and a male quick connect fitting.
  • These complete assemblies are designed specifically for use with Basic Work Bench in a laboratory environment.
  • All quick connects have 316 stainless steel internal components.
  • Keyed quick connect members will only couple with components of the same key which are identified by a color code. 
Specifications
Product Type Nitrogen Service Complete Plumbing Assembly
Color Color Coded Yellow
Material Reinforced PVC Hose, Polished Chrome Turret
Includes Male Quick Connect, System Hose and Service Turret