Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Basic Vortex Mixers

Catalog No. 88882012 Shop All Thermo Scientific Products
Change view
Click to view available options
Plug Type:
EU, UK, CN Plug
US Plug
Certifications/Compliance:
CE
cCSAus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Certifications/Compliance
88-882-012 EU, UK, CN Plug CE
88-882-011 US Plug cCSAus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 88-882-012 Supplier Thermo Scientific™ Supplier No. 88882012
  • $881.50
    $612.65 / Each of 1

    Save $268.85

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Includes

Standard package includes the Rubber platform 88882120 x 1

The Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.

The Thermo Scientific™ Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.

  • Digital Speed display
  • Continuous and Touch modes
  • Speed range: 200 to 3000rpm
  • Speed Accuracy: ± 5% rpm
  • Convenient Speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels

Order Info

Please order various accessories as the adapter for multiple applications

TRUSTED_SUSTAINABILITY

Specifications

Orbit 4 mm
Load Bearing Capacity (Metric) 0.5 kg
Speed Range 200 to 3000 rpm
Electrical Requirements 100/240 V
Voltage 100/240 V
Holds Tubes
Product Type Basic Vortex Mixer
Certifications/Compliance CE
Plug Type EU, UK, CN Plug
Frequency 50/60 Hz
Material ABS housing
Dimensions (L x W x H) 290 x 270 x 240 mm (11.4 x 10.7 x 9.4 in.)
Length (English) 11.4 in.
Length (Metric) 290 mm
Shipping Weight (English) 14.99 lb.
Shipping Weight (Metric) 6.8 kg
Weight (English) 14.33 lb.
Weight (Metric) 6.5 kg
Model Basic Vortex Mixer
Display Digital
For Use With (Equipment) Accessories: 88882120, 88882121, 88882122, 88882123, 88882124, 88882125, 88882126, 88882127
Includes Rubber platform 88882120
IP Rating IP32
Motor Type DC motor
No. of Positions 19
Operating Modes Touch, Continous
Warranty and Service Offering 2 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.