Learn More
Medchemexpress LLC Basigin/CD147 Mouse 10ug
SDP
Catalog No. 5000182426
Encompass_Preferred
The Basigin/CD147 protein is essential for retinal maturation and development acting as a NXNL1 receptor to promote the survival of retinal cone photoreceptor cells Basigin/CD147 Protein Mouse (304a a HEK293 His) is the recombinant mouse-derived Basigin/CD147 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82426 Supplier Medchemexpress LLC Supplier No. HYP7275110UG
The Basigin/CD147 protein is essential for retinal maturation and development acting as a NXNL1 receptor to promote the survival of retinal cone photoreceptor cells Basigin/CD147 Protein Mouse (304a a HEK293 His) is the recombinant mouse-derived Basigin/CD147 protein expressed by HEK293 with C-6 His labeled tag