Learn More
TARGETMOL CHEMICALS INC BASIMGLURANT 5MG
SDP
Catalog No. 502581429
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Basimglurant (CTEP Derivative) is a potent selective and orally available modulator of mGlu5 negative allosteric(Kd of 1.1 nM). purity: 99%
Catalog No. 50-258-1429 Supplier TARGETMOL CHEMICALS INC Supplier No. T127175MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Basimglurant (CTEP Derivative) is a potent selective and orally available modulator of mGlu5 negative allosteric(Kd of 1.1 nM). purity: 99%