Learn More
Med Vet International Miltex Bone Graft Basin, Holds 20 Grams, 62-71
Catalog No. 502828971
Encompass_Preferred
Stainless steel deep dish with weighted bottom to prevent tipping. Holds 20 grams of wetted material. Autoclavable.
Catalog No. 50-282-8971 Supplier Med Vet International Supplier No. MIL6271
Stainless steel deep dish with weighted bottom to prevent tipping. Holds 20 grams of wetted material. Autoclavable.