Learn More
Thomas Scientific Basins, 25mL, Sterile, Individually Wrapped, DNA-RNAse & Pyrogen Free
SDP
Catalog No. 501457352
Encompass_Preferred
Pipetting Reservoir Basins featuring Snap & Tear™ easy opening bags Designed with angled sides and a narrow V-shaped bottom to ensure that every last drop of costly reagents can be aspirated White polystyrene basins are offered in 25, 50 and 100mL volumes Thick-wall construction provides a noticeable difference in product and performance Low cross bars on each end of the basin creates much more rigidity and durability than competitive offerings Pour spouts in every corner, and one larger spout on each reservoir to facilitate spill-free pouring Individually wrapped and 5-pack versions are packaged in easy-open bags - Snap & Tear! No need for scissors or concerns about damaging the basins as you open the bags
This item is not returnable. View return policy
Catalog No. 50-145-7352 Supplier Thomas Scientific Supplier No. 1185A00
Pipetting Reservoir Basins featuring Snap & Tear™ easy opening bags Designed with angled sides and a narrow V-shaped bottom to ensure that every last drop of costly reagents can be aspirated White polystyrene basins are offered in 25, 50 and 100mL volumes Thick-wall construction provides a noticeable difference in product and performance Low cross bars on each end of the basin creates much more rigidity and durability than competitive offerings Pour spouts in every corner, and one larger spout on each reservoir to facilitate spill-free pouring Individually wrapped and 5-pack versions are packaged in easy-open bags - Snap & Tear! No need for scissors or concerns about damaging the basins as you open the bags