Learn More
Basix™ Empty Racks with Inserts
GREENER_CHOICE
Catalog No. 13611100 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
200μL tips
Catalog No. For Use With (Equipment)
13-611-100 200μL tips
1 options
Catalog No. 13-611-100
Add to Cart
Add to Cart
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everdayliquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Sterility Non-sterile
Autoclavable Autoclavable
For Use With (Equipment) 200μL tips
Material Polypropylene