Learn More
World Precision Instrument Sterilization Baskets, Side Perforated, Flat Base-WP-4547
SDP
Catalog No. 502090428
Encompass
Stainless steel Sterilization Baskets for Instrument cleaning, sterilization and storage.  The perforated side walls protect instrument edges and tips from damage.
These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Catalog No. 50-209-0428 Supplier World Precision Instrument Supplier No. WP4547
Stainless steel Sterilization Baskets for Instrument cleaning, sterilization and storage.  The perforated side walls protect instrument edges and tips from damage.
These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.