Learn More
World Precision Instrument Sterilization Baskets, Side Perforated, 240 x 250 x 100 mm, Flat Base-WP-4552
SDP
Catalog No. 502090433
Encompass
Stainless steel Sterilization Baskets for Instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage.
These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Catalog No. 50-209-0433 Supplier World Precision Instrument Supplier No. WP4552
Stainless steel Sterilization Baskets for Instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage.
These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.