Learn More
ELITechGroup Aerospray Hematology Stat Reagent C, Basofix™ Eosin Stain, 500mL
SDP
Catalog No. NC9137907
Encompass
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Red; 6.65 +/- 0.05 pH; Hematology Reagent C Basofix Eosin Stain; B.P. ~100 deg.C; Freezing Point ~0 deg.C; Storage: RT away from direct light
Catalog No. NC9137907 Supplier ELITechGroup Supplier No. SS049C
This item has a minimum order quantity of 4 per supplier requirements. Aqueous Eosin Y Stain; Liquid, Red; 6.65 +/- 0.05 pH; Hematology Reagent C Basofix Eosin Stain; B.P. ~100 deg.C; Freezing Point ~0 deg.C; Storage: RT away from direct light