Learn More
ELITechGroup Aerospray Hematology Stat, Basofix™ Pre-dip Fixative, 500mL
SDP
Catalog No. NC9137908
Encompass
Basofix is a system of reagents for staining peripheral blood smears with the Wescor Aerospray Hematology Slide Stainer. Its purpose is to fix granules of basophilic leukocytes in the slide.
Catalog No. NC9137908 Supplier ELITechGroup Supplier No. SS049P
Basofix is a system of reagents for staining peripheral blood smears with the Wescor Aerospray Hematology Slide Stainer. Its purpose is to fix granules of basophilic leukocytes in the slide.